Assay Details
| Gene Symbol: Rasa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039103.3 | NP_001034192.1 | 18 - 19 | 2283 | 96 | |
| NM_133914.3 | NP_598675.2 | 19 - 20 | 2421 | 96 | ||
| GenBank mRNA | AK014220.1 | - | 18 - 19 | 2570 | 96 | |
| AK154335.1 | - | 19 - 20 | 2422 | 96 | ||
| AK155146.1 | - | 19 - 20 | 2422 | 96 | ||
| AK220218.1 | - | 19 - 20 | 2297 | 96 | ||
| AY591339.1 | - | 19 - 20 | 2322 | 96 | ||
| BC027230.1 | - | 18 - 19 | 2542 | 96 | ||
| BC049381.1 | - | 18 - 19 | 2916 | 96 | ||
| BC057460.1 | - | 19 - 20 | 2275 | 4527934 | 96 | |
| BC092143.1 | - | 1 - 2 | 105 | 96 | ||
| DQ317932.1 | - | 18 - 19 | 2167 | 96 | ||
More Information
Set Membership: |
|

Rasa4
Genomic Map