Assay Details
| Gene Symbol: Mobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039364.2 | NP_001034453.1 | 1 - 2 | 51 | 65 | |
| GenBank mRNA | BY257495.1 | - | 1 - 2 | 52 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001039364.2 transcripts.
Set Membership: |
|

Mobp
Genomic Map