Assay Details
| Gene Symbol: Uqcr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025650.2 | NP_079926.1 | 1 - 2 | 124 | 61 | |
| GenBank mRNA | AK003033.1 | - | 1 - 2 | 134 | 61 | |
| AK003443.1 | - | 1 - 2 | 104 | 61 | ||
| AK013513.1 | - | 1 - 2 | 72 | 61 | ||
| AK167946.1 | - | 1 - 2 | 120 | 61 | ||
| AK168974.1 | - | 1 - 2 | 89 | 61 | ||
| BC034408.1 | - | 1 - 2 | 88 | 4235605 | 61 | |
| BU529273.1 | - | 1 - 2 | 125 | 61 | ||
More Information
Set Membership: |
|

Uqcr11
Genomic Map