Assay Details
| Gene Symbol: Cdca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025866.3 | NP_080142.1 | 3 - 4 | 480 | 93 | |
| GenBank mRNA | AK011289.1 | - | 3 - 4 | 481 | 93 | |
| AK028671.1 | - | 3 - 4 | 491 | 93 | ||
| AK075882.1 | - | 3 - 4 | 445 | 93 | ||
| AK153943.1 | - | 3 - 4 | 459 | 93 | ||
| AK167734.1 | - | 3 - 4 | 481 | 93 | ||
| BC066169.1 | - | 3 - 4 | 488 | 30474357 | 93 | |
More Information
Set Membership: |
|

Cdca7
Genomic Map