Assay Details
Gene Symbol: Higd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025933.3 | NP_080209.1 | 1 - 2 | 196 | 75 | |
GenBank mRNA | AK004272.1 | - | 1 - 2 | 36 | 75 | |
AK005269.1 | - | 1 - 2 | 197 | 75 | ||
AK008387.1 | - | 1 - 2 | 193 | 75 | ||
BC021471.1 | - | 1 - 2 | 193 | 5343356 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
