Assay Details
| Gene Symbol: Mrpl42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026065.3 | NP_080341.1 | 5 - 6 | 493 | 68 | |
| GenBank mRNA | AK002530.1 | - | 5 - 6 | 478 | 68 | |
| AK003427.2 | - | 5 - 6 | 494 | 68 | ||
| AK013693.1 | - | 5 - 6 | 490 | 68 | ||
| BC024337.1 | - | 5 - 6 | 474 | 4984291 | 68 | |
More Information
Set Membership: |
|

Mrpl42
Genomic Map