Assay Details
| Gene Symbol: Mrpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026154.1 | NP_080430.1 | 2 - 3 | 235 | 96 | |
| GenBank mRNA | AK002865.1 | - | 2 - 3 | 235 | 96 | |
| AK007643.1 | - | 2 - 3 | 227 | 96 | ||
| AK150774.1 | - | 2 - 3 | 227 | 96 | ||
| AK171028.1 | - | 2 - 3 | 229 | 96 | ||
| BC016219.1 | - | 2 - 3 | 221 | 4485332 | 96 | |
More Information
Set Membership: |
|

Mrpl10
Genomic Map