Assay Details
| Gene Symbol: Aasdhppt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026276.3 | NP_080552.3 | 2 - 3 | 467 | 109 | |
| GenBank mRNA | AK007737.1 | - | 2 - 3 | 468 | 109 | |
| AK010557.1 | - | 2 - 3 | 427 | 109 | ||
| AK013111.1 | - | 2 - 3 | 421 | 109 | ||
| AK014188.1 | - | 2 - 3 | 410 | 109 | ||
| AK018191.1 | - | 1 - 2 | 113 | 109 | ||
| AK028700.1 | - | 2 - 3 | 424 | 109 | ||
| AK029523.1 | - | 2 - 3 | 409 | 109 | ||
| AK076099.1 | - | 2 - 3 | 422 | 109 | ||
| AK076866.1 | - | 2 - 3 | 441 | 109 | ||
| AK081818.1 | - | 2 - 3 | 443 | 109 | ||
| AK144671.1 | - | 2 - 3 | 441 | 109 | ||
| BC030043.1 | - | 2 - 3 | 402 | 1348651 | 109 | |
| BC038013.1 | - | 2 - 3 | 438 | 3497754 | 109 | |
| BC049851.1 | - | 2 - 3 | 428 | 3481972 | 109 | |
More Information
Set Membership: |
|

Aasdhppt
Genomic Map