Assay Details
| Gene Symbol: Ppid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026352.3 | NP_080628.1 | 6 - 7 | 836 | 109 | |
| GenBank mRNA | AK003402.1 | - | 6 - 7 | 793 | 109 | |
| AK013919.1 | - | 6 - 7 | 827 | 109 | ||
| AK051597.1 | - | 6 - 7 | 816 | 109 | ||
| AK142413.1 | - | 5 - 6 | 3260 | 109 | ||
| BC011499.1 | - | 6 - 7 | 833 | 3584183 | 109 | |
| BC019778.1 | - | 6 - 7 | 836 | 4035785 | 109 | |
More Information
Set Membership: |
|

Ppid
Genomic Map