Assay Details
| Gene Symbol: Tceb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026456.4 | NP_080732.1 | 1 - 2 | 34 | 102 | |
| GenBank mRNA | AK011958.1 | - | 1 - 2 | 54 | 102 | |
| AK012909.1 | - | 1 - 2 | 35 | 102 | ||
| AK013089.1 | - | 1 - 2 | 51 | 102 | ||
| AK039746.1 | - | 1 - 2 | 35 | 102 | ||
| BC009104.1 | - | 1 - 2 | 34 | 3483215 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011958.1, AK012909.1, AK013089.1, AK039746.1, BC009104.1, NM_026456.4 transcripts.
Set Membership: |
|

Tceb1
Genomic Map