Assay Details
Gene Symbol: Mphosph6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026758.3 | NP_081034.1 | 1 - 2 | 83 | 91 | |
GenBank mRNA | AK003245.1 | - | 1 - 2 | 84 | 91 | |
BC008161.1 | - | 1 - 2 | 79 | 3964232 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
