Assay Details
| Gene Symbol: Hexim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130515.1 | NP_001123987.1 | 2 - 3 | 180 | 114 | |
| NM_001130516.1 | NP_001123988.1 | 2 - 3 | 197 | 114 | ||
| NM_027658.2 | NP_081934.1 | 2 - 3 | 209 | 114 | ||
| GenBank mRNA | AK016624.1 | - | 2 - 3 | 220 | 114 | |
| AK140145.1 | - | 1 - 2 | 108 | 114 | ||
| AK160110.1 | - | 2 - 3 | 198 | 114 | ||
| BC026458.1 | - | 2 - 3 | 205 | 4481287 | 114 | |
| BC115597.1 | - | 2 - 3 | 173 | 40102543 | 114 | |
| BC115598.1 | - | 2 - 3 | 173 | 40102544 | 114 | |
| BY295186.1 | - | 2 - 3 | 210 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016624.1 transcripts.
Set Membership: |
|

Hexim2
Genomic Map