Assay Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027727.2 | NP_082003.1 | 11 - 12 | 1512 | 99 | |
| GenBank mRNA | AK049909.1 | - | 11 - 12 | 1477 | 99 | |
| AK140643.1 | - | 3 - 4 | 617 | 99 | ||
| AK154794.1 | - | 11 - 12 | 1501 | 99 | ||
| BC018220.1 | - | 7 - 8 | 872 | 3985017 | 99 | |
| BC038276.1 | - | 11 - 12 | 1514 | 4458792 | 99 | |
More Information
Set Membership: |
|

Dym
Genomic Map