Assay Details
| Gene Symbol: Cldn34c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028426.1 | NP_082702.1 | 3 - 4 | 235 | 62 | |
| GenBank mRNA | AK014019.1 | - | 3 - 4 | 203 | 62 | |
| BC027572.1 | - | 3 - 4 | 235 | 1363266 | 62 | |
More Information
Set Membership: |
|

Cldn34c1
Genomic Map