Assay Details
Gene Symbol: Gpbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029868.2 | NP_084144.2 | 6 - 7 | 1711 | 66 | |
GenBank mRNA | AK037224.1 | - | 6 - 7 | 1732 | 66 | |
AK077148.1 | - | 6 - 7 | 1681 | 66 | ||
AK133761.1 | - | 6 - 7 | 1727 | 66 | ||
BC031428.1 | - | 5 - 6 | 784 | 4021073 | 66 | |
BC066034.1 | - | 6 - 7 | 1711 | 6836605 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
