Assay Details
| Gene Symbol: Slc35e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029875.2 | NP_084151.2 | 4 - 5 | 923 | 80 | |
| GenBank mRNA | AK145213.1 | - | 4 - 5 | 416 | 80 | |
| BC006601.1 | - | 4 - 5 | 942 | 3965585 | 80 | |
| BC057101.1 | - | 4 - 5 | 923 | 5720169 | 80 | |
More Information
Set Membership: |
|

Slc35e3
Genomic Map