Assay Details
| Gene Symbol: Nol3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030152.4 | NP_084428.1 | 2 - 3 | 469 | 97 | |
| GenBank mRNA | AK021023.1 | - | 2 - 3 | 415 | 97 | |
| AK053846.1 | - | 2 - 3 | 470 | 97 | ||
| AK079536.1 | - | 2 - 3 | 501 | 97 | ||
| AY459322.1 | - | 2 - 3 | 477 | 97 | ||
| BC027290.1 | - | 2 - 3 | 496 | 2647023 | 97 | |
| BC053971.1 | - | 2 - 3 | 441 | 97 | ||
More Information
Set Membership: |
|

Nol3
Genomic Map