Assay Details
| Gene Symbol: Ier5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030244.3 | NP_084520.3 | 1 | 921 | 64 | |
| GenBank mRNA | AK012145.1 | - | 1 | 935 | 64 | |
| AK034055.1 | - | 944 | 64 | |||
| BC003464.1 | - | 1 | 920 | 2648900 | 64 | |
| BC010234.1 | - | 1 | 921 | 3499467 | 64 | |
| BC040084.1 | - | 1 | 921 | 3499467 | 64 | |
More Information
Set Membership: |
|

Ier5l
Genomic Map