Assay Details
| Gene Symbol: Laptm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033521.3 | NP_277056.1 | 1 - 2 | 402 | 84 | |
| GenBank mRNA | AF317418.1 | - | 1 - 2 | 104 | 84 | |
| AK077269.1 | - | 1 - 2 | 265 | 84 | ||
| AK077933.1 | - | 1 - 2 | 402 | 84 | ||
| AK079581.1 | - | 1 - 2 | 265 | 84 | ||
| AK082807.1 | - | 1 - 2 | 281 | 84 | ||
| AK166014.1 | - | 1 - 2 | 304 | 84 | ||
| BC019120.1 | - | 1 - 2 | 272 | 4507607 | 84 | |
| EU561010.1 | - | 1 - 2 | 402 | 84 | ||
More Information
Set Membership: |
|

Laptm4b
Genomic Map