Assay Details
Gene Symbol: Cox6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053071.2 | NP_444301.1 | 2 - 3 | 256 | 74 | |
GenBank mRNA | AK012602.1 | - | 2 - 3 | 173 | 74 | |
AK013459.1 | - | 2 - 3 | 172 | 74 | ||
BC024666.1 | - | 2 - 3 | 187 | 4195595 | 74 | |
BC094413.1 | - | 2 - 3 | 159 | 1397127 | 74 | |
BY100624.1 | - | 2 - 3 | 258 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
