Assay Details
| Gene Symbol: Osr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054049.2 | NP_473390.1 | 3 - 4 | 1228 | 65 | |
| GenBank mRNA | AF370121.1 | - | 3 - 4 | 1228 | 65 | |
| AK052147.1 | - | 3 - 4 | 1243 | 65 | ||
| BC013504.1 | - | 3 - 4 | 1035 | 4224339 | 65 | |
More Information
Set Membership: |
|

Osr2
Genomic Map