Assay Details
| Gene Symbol: Cyp4v3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133969.2 | NP_598730.1 | 1 - 2 | 431 | 85 | |
| GenBank mRNA | AB056457.1 | - | 1 - 2 | 397 | 85 | |
| AK004724.1 | - | 1 - 2 | 431 | 85 | ||
| AK150653.1 | - | 1 - 2 | 431 | 85 | ||
| AK165754.1 | - | 397 | 85 | |||
| BC138967.1 | - | 1 - 2 | 297 | 8861989 | 85 | |
| BC138968.1 | - | 1 - 2 | 297 | 8861990 | 85 | |
More Information
Set Membership: |
|

Cyp4v3
Genomic Map