Assay Details
| Gene Symbol: Trim41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145377.2 | NP_663352.2 | 2 - 3 | 1646 | 86 | |
| GenBank mRNA | BC041780.1 | - | 2 - 3 | 1608 | 86 | |
More Information
Set Membership: |
|

Trim41
Genomic Map