Assay Details
| Gene Symbol: L2hgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145443.2 | NP_663418.1 | 4 - 5 | 617 | 122 | |
| GenBank mRNA | AK145228.1 | - | 4 - 5 | 616 | 122 | |
| AK152450.1 | - | 4 - 5 | 594 | 122 | ||
| AK168429.1 | - | 4 - 5 | 565 | 122 | ||
| BC016226.1 | - | 4 - 5 | 561 | 4487011 | 122 | |
More Information
Set Membership: |
|

L2hgdh
Genomic Map