Assay Details
| Gene Symbol: Gnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145552.2 | NP_663527.2 | 15 - 16 | 2158 | 103 | |
| GenBank mRNA | AK077532.1 | - | 15 - 16 | 2128 | 103 | |
| AK146404.1 | - | 15 - 16 | 2155 | 103 | ||
| BC003262.1 | - | 15 - 16 | 2134 | 3501393 | 103 | |
More Information
Set Membership: |
|

Gnl2
Genomic Map