Assay Details
| Gene Symbol: Efemp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146015.2 | NP_666127.2 | 10 - 11 | 1454 | 128 | |
| GenBank mRNA | AK077302.1 | - | 10 - 11 | 1456 | 128 | |
| AY185605.1 | - | 9 - 10 | 1279 | 128 | ||
| AY251056.1 | - | 10 - 11 | 1449 | 128 | ||
| BC023060.1 | - | 3 - 4 | 410 | 128 | ||
| BC031184.1 | - | 7 - 8 | 1075 | 4989603 | 128 | |
More Information
Set Membership: |
|

Efemp1
Genomic Map