Assay Details
Gene Symbol: Olfr30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146878.2 | NP_667089.1 | 1 | 518 | 86 | |
GenBank mRNA | AY043304.1 | - | 1 | 102 | 86 | |
BC119375.1 | - | 1 | 518 | 8734124 | 86 | |
X89680.1 | - | 1 | 108 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
