Assay Details
| Gene Symbol: Wasf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153423.6 | NP_700472.1 | 1 - 2 | 92 | 90 | |
| GenBank mRNA | AK085458.1 | - | 1 - 2 | 93 | 90 | |
| AK150140.1 | - | 1 - 2 | 161 | 90 | ||
| AK172187.1 | - | 1 - 2 | 124 | 90 | ||
| AK183500.1 | - | 1 - 2 | 43 | 91 | ||
| BC051488.1 | - | 1 - 2 | 156 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK085458.1, AK172187.1, BC051488.1, NM_153423.6 transcripts.
Set Membership: |
|

Wasf2
Genomic Map