Assay Details
| Gene Symbol: Nars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153591.4 | NP_705819.3 | 3 - 4 | 651 | 81 | |
| GenBank mRNA | AK046799.1 | - | 3 - 4 | 652 | 81 | |
| AK046801.1 | - | 3 - 4 | 652 | 81 | ||
| AK047091.1 | - | 3 - 4 | 596 | 81 | ||
| AK050718.1 | - | 3 - 4 | 590 | 81 | ||
| AK138260.1 | - | 3 - 4 | 631 | 81 | ||
More Information
Set Membership: |
|

Nars2
Genomic Map