Assay Details
| Gene Symbol: Aunip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081099.1 | NP_001074568.1 | 1 - 2 | 169 | 62 | |
| GenBank mRNA | AK011279.1 | - | 1 - 2 | 94 | 62 | |
| BC138271.1 | - | 1 - 2 | 145 | 8861290 | 62 | |
More Information
Set Membership: |
|

Aunip
Genomic Map