Assay Details
| Gene Symbol: Stk38l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172734.3 | NP_766322.1 | 2 - 3 | 262 | 93 | |
| GenBank mRNA | AK052288.1 | - | 2 - 3 | 198 | 93 | |
| AY223819.1 | - | 2 - 3 | 184 | 93 | ||
| BC062170.1 | - | 1 - 2 | 156 | 30135051 | 93 | |
| CJ082246.1 | - | 2 - 3 | 262 | 93 | ||
More Information
Set Membership: |
|

Stk38l
Genomic Map