Assay Details
| Gene Symbol: Stk38l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172734.3 | NP_766322.1 | 3 - 4 | 308 | 76 | |
| GenBank mRNA | AK052288.1 | - | 3 - 4 | 244 | 76 | |
| AY223819.1 | - | 3 - 4 | 230 | 76 | ||
| BC062170.1 | - | 2 - 3 | 202 | 30135051 | 76 | |
| CJ082246.1 | - | 3 - 4 | 308 | 76 | ||
More Information
Set Membership: |
|

Stk38l
Genomic Map