Assay Details
| Gene Symbol: Idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173011.2 | NP_766599.2 | 3 - 4 | 442 | 89 | |
| GenBank mRNA | AF212319.1 | - | 3 - 4 | 413 | 89 | |
| AK088110.1 | - | 3 - 4 | 414 | 89 | ||
| AK145753.1 | - | 3 - 4 | 387 | 89 | ||
| AK146564.1 | - | 3 - 4 | 425 | 89 | ||
| AK161640.1 | - | 3 - 4 | 426 | 89 | ||
| AK169395.1 | - | 3 - 4 | 439 | 89 | ||
| AK212860.1 | - | 1 - 2 | 147 | 89 | ||
| BC060030.1 | - | 3 - 4 | 430 | 6515657 | 89 | |
More Information
Set Membership: |
|

Idh2
Genomic Map