Assay Details
| Gene Symbol: Lrit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173418.3 | NP_775594.1 | 1 - 2 | 139 | 87 | |
| GenBank mRNA | AK044383.1 | - | 1 - 2 | 140 | 87 | |
| AK044611.1 | - | 1 - 2 | 141 | 87 | ||
| BC057628.1 | - | 1 - 2 | 114 | 4511498 | 87 | |
More Information
Set Membership: |
|

Lrit2
Genomic Map