Assay Details
| Gene Symbol: Zfp770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175466.4 | NP_780675.1 | 1 - 2 | 103 | 83 | |
| GenBank mRNA | AK032578.1 | - | 1 - 2 | 104 | 83 | |
| AK043843.1 | - | 1 - 2 | 141 | 83 | ||
| AK077547.1 | - | 1 - 2 | 104 | 83 | ||
| AK164226.1 | - | 1 - 2 | 86 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032578.1, AK043843.1, AK077547.1, AK164226.1, NM_175466.4 transcripts.
Set Membership: |
|

Zfp770
Genomic Map