Assay Details
| Gene Symbol: Acer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175731.4 | NP_783858.1 | 2 - 3 | 253 | 96 | |
| GenBank mRNA | AF347023.1 | - | 2 - 3 | 235 | 96 | |
| AK028901.1 | - | 2 - 3 | 254 | 96 | ||
| AK075884.1 | - | 2 - 3 | 239 | 96 | ||
| BC130254.1 | - | 2 - 3 | 258 | 40142524 | 96 | |
More Information
Set Membership: |
|

Acer1
Genomic Map