Assay Details
| Gene Symbol: Gchfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177157.4 | NP_796131.1 | 1 - 2 | 86 | 81 | |
| GenBank mRNA | AK028116.1 | - | 1 - 2 | 87 | 81 | |
| AK028122.1 | - | 1 - 2 | 87 | 81 | ||
| BC096406.1 | - | 1 - 2 | 74 | 1511037 | 81 | |
| BC132450.1 | - | 1 - 2 | 82 | 40130727 | 81 | |
| BC132478.1 | - | 1 - 2 | 82 | 40130755 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028116.1, AK028122.1, BC096406.1, BC132450.1, BC132478.1, NM_177157.4 transcripts.
Set Membership: |
|

Gchfr
Genomic Map