Assay Details
| Gene Symbol: Zfp281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177643.4 | NP_808311.1 | 1 - 2 | 153 | 95 | |
| GenBank mRNA | AK142218.1 | - | 1 - 2 | 70 | 95 | |
| AK157189.1 | - | 1 - 2 | 96 | 95 | ||
| AV581959.1 | - | 1 - 2 | 186 | 95 | ||
| BC003243.1 | - | 1 - 2 | 114 | 3498439 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK142218.1, AK157189.1, BC003243.1, NM_177643.4 transcripts.
Set Membership: |
|

Zfp281
Genomic Map