Assay Details
| Gene Symbol: Gsdmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168274.1 | NP_001161746.1 | 1131 | 108 | ||
| NM_177912.4 | NP_808580.2 | 896 | 108 | |||
| GenBank mRNA | AK033603.1 | - | 897 | 108 | ||
| BC113153.1 | - | 889 | 40058717 | 108 | ||
| BC113155.1 | - | 1117 | 40058514 | 108 | ||
More Information
Set Membership: |
|

Gsdmc2
Genomic Map