Assay Details
| Gene Symbol: Hist1h4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178192.2 | NP_835499.1 | 1 | 421 | 65 | |
| GenBank mRNA | AK016310.1 | - | 1 | 403 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h4a| Gene Symbol: Hist1h4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178192.2 | NP_835499.1 | 1 | 421 | 65 | |
| GenBank mRNA | AK016310.1 | - | 1 | 403 | 65 | |
Set Membership: |
|