Assay Details
| Gene Symbol: Zrsr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009453.3 | NP_033479.2 | 4 - 5 | 657 | 140 | |
| NM_178794.4 | NP_848909.2 | 4 - 5 | 657 | 140 | ||
| GenBank mRNA | AK030296.1 | - | 4 - 5 | 589 | 140 | |
| AK045504.1 | - | 4 - 5 | 589 | 140 | ||
| AK078070.1 | - | 4 - 5 | 302 | 140 | ||
| BC065693.1 | - | 4 - 5 | 573 | 140 | ||
| BC083160.1 | - | 4 - 5 | 550 | 140 | ||
| BC099617.1 | - | 4 - 5 | 555 | 140 | ||
| D45205.1 | - | 4 - 5 | 580 | 140 | ||
More Information
Set Membership: |
|

Zrsr2
Genomic Map