Assay Details
| Gene Symbol: Ccnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183015.3 | NP_898836.2 | 11 - 12 | 3959 | 92 | |
| GenBank mRNA | AJ416459.1 | - | 3 - 4 | 356 | 92 | |
| AJ555464.1 | - | 11 - 12 | 3960 | 92 | ||
| AJ555465.1 | - | 10 - 11 | 1235 | 92 | ||
| BC132411.1 | - | 10 - 11 | 3913 | 40130688 | 92 | |
| BC138355.1 | - | 10 - 11 | 3913 | 8861375 | 92 | |
More Information
Set Membership: |
|

Ccnb3
Genomic Map