Assay Details
| Gene Symbol: Cct8l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198621.2 | NP_941023.1 | 1 | 1625 | 66 | |
| GenBank mRNA | AK133375.1 | - | 1 | 1625 | 66 | |
| BC050797.1 | - | 1 | 1542 | 6743432 | 66 | |
More Information
Set Membership: |
|

Cct8l1
Genomic Map