Assay Details
Gene Symbol: Tas2r107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC141027.1 | - | 1 | 823 | 9055639 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tas2r107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC141027.1 | - | 1 | 823 | 9055639 | 89 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |