Assay Details
| Gene Symbol: Krt78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212487.4 | NP_997652.4 | 6 - 7 | 1089 | 63 | |
| GenBank mRNA | AK146611.1 | - | 6 - 7 | 1086 | 63 | |
| AY651827.1 | - | 6 - 7 | 914 | 63 | ||
| BC119600.1 | - | 6 - 7 | 1026 | 63 | ||
| BC119601.1 | - | 6 - 7 | 1026 | 63 | ||
| BC127241.1 | - | 6 - 7 | 1026 | 63 | ||
| BC127618.1 | - | 6 - 7 | 1026 | 63 | ||
More Information
Set Membership: |
|

Krt78
Genomic Map