Assay Details
| Gene Symbol: Cd72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110321.1 | NP_001103791.1 | 2 - 3 | 281 | 99 | |
| GenBank mRNA | AK144191.1 | - | 2 - 3 | 284 | 99 | |
More Information
Set Membership: |
|

Cd72
Genomic Map