Assay Details
| Gene Symbol: Myl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113387.1 | NP_001106858.1 | 5 - 6 | 507 | 91 | |
| NM_021285.3 | NP_067260.1 | 5 - 6 | 684 | 91 | ||
| GenBank mRNA | AK003182.1 | - | 5 - 6 | 660 | 91 | |
| AK009938.1 | - | 5 - 6 | 496 | 91 | ||
| W48198.1 | - | 2 - 3 | 190 | 91 | ||
More Information
Set Membership: |
|

Myl1
Genomic Map