Assay Details
Gene Symbol: Nfe2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130454.1 | NP_001123926.1 | 1 - 2 | 69 | 87 | |
GenBank mRNA | AK148471.1 | - | 1 - 2 | 69 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
