Assay Details
| Gene Symbol: Hexim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130516.1 | NP_001123988.1 | 1 - 2 | 80 | 107 | |
| GenBank mRNA | AK160110.1 | - | 1 - 2 | 81 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK160110.1, NM_001130516.1 transcripts.
Set Membership: |
|

Hexim2
Genomic Map