Assay Details
| Gene Symbol: Lrrc74b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145435.1 | NP_001138907.1 | 4 - 5 | 653 | 81 | |
| NM_029053.3 | NP_083329.2 | 4 - 5 | 653 | 81 | ||
| GenBank mRNA | AK039092.1 | - | 4 - 5 | 631 | 81 | |
| AK132588.1 | - | 4 - 5 | 651 | 81 | ||
| AK143245.1 | - | 4 - 5 | 626 | 81 | ||
| AK161226.1 | - | 4 - 5 | 651 | 81 | ||
| BC115678.1 | - | 4 - 5 | 630 | 40104166 | 81 | |
| BC115679.1 | - | 4 - 5 | 630 | 40104167 | 81 | |
More Information
Set Membership: |
|

Lrrc74b
Genomic Map